Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
In Western blots, 100% cross-reactivity with recombinant human (rh) Themis (aa 2-282) is observed under non-reducing conditions, and approximately 10% cross-reactivity with rhThemis (aa 2-282) is observed under reducing conditions.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA325O24.3; bA325O24.4; FLJ40584; Gasp; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; Thymocyte-expressed molecule involved in selection
Gene Aliases: E430004N04Rik; Gasp; Spot; Themis; thylex; Tsepa
UniProt ID: (Mouse) Q8BGW0
Entrez Gene ID: (Mouse) 210757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support