Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA5-14705 targets Thyroid Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples.
The MA5-14705 immunogen is human Thyroid Stimulating Hormone.
MA5-14705 detects Thyroid Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa.
Product MA514705 is a smaller package size of MIT0401 (formerly sold as a Seradyn product).
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anterior pituitary glycoprotein hormones common subunit alpha; CG-alpha; Choriogonadotropin alpha chain; Chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; Follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; Follitropin alpha chain; FSH-alpha; Glycoprotein hormones alpha chain; LSH-alpha; Luteinizing hormone alpha chain; Lutropin alpha chain; thyroid stimulating hormone, beta; Thyroid-stimulating hormone alpha chain; Thyroid-stimulating hormone subunit beta; Thyrotropin; Thyrotropin alpha chain; Thyrotropin beta chain; Thyrotropin subunit beta; TSH-alpha; TSH-B
Gene Aliases: CG-ALPHA; CGA; FSHA; GPHa; GPHA1; HCG; LHA; TSH-B; TSH-BETA; TSHA; TSHB
UniProt ID: (Human) P01215, (Human) P01222
Entrez Gene ID: (Human) 1081, (Human) 7252
Molecular Function:
peptide hormone
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support