Invitrogen
MA5-14688 targets Thyroid Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples.
The MA5-14688 immunogen is human Thyroid Stimulating Hormone.
MA5-14688 detects Thyroid Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa.
Product MA514688 is a smaller package size of MIT0412 (formerly sold as a Seradyn product).
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.