Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.^M
^M
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: formin binding protein 1-like; Formin-binding protein 1-like; Toca-1; transducer of Cdc42-dependent actin assembly 1; Transducer of Cdc42-dependent actin assembly protein 1
Gene Aliases: C1orf39; FNBP1L; TOCA1
UniProt ID: (Human) Q5T0N5
Entrez Gene ID: (Human) 54874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support