Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive test controls include: 293T, HepG2, U-87MG, Mouse liver, Mouse brain, Mouse spleen, Rat lung.
Immunogen sequence: LKDIEHALSV SVFNNKNSGF WHSSLIDRNL IDYFVPFLPL EYKHLKMCIR VEMQSRGYEI DEDIVSRVAE EMTFFPKEER VFSDKGCKTV FTKLDYYYDD
The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); Torsin 1A; torsin A; Torsin ATPase 1; Torsin ATPase-1A; Torsin family 1 member A; torsin family 1, member A (torsin A); Torsin-1A
Gene Aliases: DQ2; DYT1; TA; TOR1A; TORA; torsinA
UniProt ID: (Human) O14656, (Rat) Q68G38, (Mouse) Q9ER39
Entrez Gene ID: (Human) 1861, (Rat) 266606, (Mouse) 30931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support