Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TOR1B may serve as a molecular chaperone assisting in the proper folding of secreted and/or membrane proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Torsin ATPase-1B; torsin B; Torsin family 1 member B; torsin family 1, member B (torsin B); Torsin-1B
Gene Aliases: 2610016F05Rik; DQ1; FKSG18; TOR1B; torsinB
UniProt ID: (Human) O14657, (Mouse) Q9ER41
Entrez Gene ID: (Human) 27348, (Rat) 311854, (Mouse) 30934
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support