Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AbboMax
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Positive control: MCF-7 cells
Cellular location: Cytoplasmic, Membrane.
It belongs to the TPD52 family. Its exact function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mD52; prostate and colon associated protein; prostate leucine zipper; Protein N8; Tumor protein D52
Gene Aliases: AI043038; D52; hD52; mD52; N8L; PC-1; PrLZ; TPD52
UniProt ID: (Human) P55327, (Mouse) Q62393
Entrez Gene ID: (Chicken) 770339, (Bovine) 540235, (Human) 7163, (Rat) 294900, (Mouse) 21985
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support