Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Suggested positive controls for this product are KG1 cells and human colorectal carcinoma tissues.
Transferrins (Tf) are an approximately 80kDa blood plasma glycoprotein synthesized by the liver. They are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate. It is responsible for the transport of iron from sites of absorption and heme degradation to those of storage and utilization. Transferrin is the primary blood iron transport protein and under normal conditions; approximately one-third of total blood transferrin contains bound iron. Measurement of blood transferrin levels can be used as an indicator for blood iron-carrying capacity and abnormalities of iron metabolism such as anaemia, iron overload and haemochromatosis.Serum transferrin may also have a further role in stimulating cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1 metal-binding globulin; DKFZp781D0156; epididymis secretory sperm binding protein Li 71p; Liver regeneration-related protein LRRG03; Serotransferrin; Siderophilin; transferin; Transferrin
Gene Aliases: HEL-S-71p; PRO1400; PRO1557; PRO2086; TF; Tfn; TFQTL1; Trf
UniProt ID: (Human) P02787, (Rat) P12346
Entrez Gene ID: (Human) 7018, (Rat) 24825
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support