Invitrogen
PA1-4111 detects mouse mammary tumor virus receptor in mouse samples.
PA1-4111 has been successfully used in Western blot procedures.
The PA1-4111 immunogen is a synthetic peptide corresponding to residues D(152) D E E P P D A S L P P C(163) of the 20 kDa MMTV-R.
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.