Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 7
This gene encodes a neuronal-specific member of the tropomodulin family of actin-regulatory proteins. The encoded protein caps the pointed end of actin filaments preventing both elongation and depolymerization. The capping activity of this protein is dependent on its association with tropomyosin. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-Tmod; neural tropomodulin; Neuronal tropomodulin; tropomodulin 2 (neuronal); Tropomodulin-2
Gene Aliases: N-TMOD; NTMOD; TMOD2
UniProt ID: (Human) Q9NZR1, (Mouse) Q9JKK7, (Rat) P70566
Entrez Gene ID: (Human) 29767, (Mouse) 50876, (Rat) 58814
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support