Search Thermo Fisher Scientific
AbboMax
FIGURE: 1 / 1
Positive control: Breast Carcinoma
Cellular location: Cytoplasmic.
Tumor-associated glycoprotein (TAG-72) is an oncofetal antigen with properties of a mucin. About 60% of carcinoma patients express TAG-72 in their sera.