Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVRPGDQQP RPLEALNGGF NLSRILRTAL PAWHKSIVSF RQLREEVLGE LSNVEQAAGL RWSRFPDLNR ILKGHRKGEL TVFTGPTGSG KTTFISEYAL DLCSQGVNTL WGSFEISNVR LARVMLTQFA EGRLEDQLDK YDHWADRFED LPLYFMTFHG QQSIRTVIDT MQHAVYVYDI CHVIIDNLQF MMGHEQLSTD RIAAQDYIIG VFRKFATDNN CHVTLVIHPR KEDDDKELQT ASIFGSAKAS QEADNVLILQ DRKLVTGPGK RYLQVSKNRF DGDVGVFPLE FNKNSLTFSI PPKNKARLKK IKDDTGPVAK KPSSGKKGAT TQNSEICSGQ APTPDQPDTS KRSK (334-684 aa encoded by B C033762)
Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars.Twinkle is a mitochondrial protein with structural similarity to the phage T7 primase/helicase (GP4) and other hexameric ring helicases. The twinkle protein colocalizes with mtDNA in mitochondrial nucleoids, and its name derives from the unusual localization pattern reminiscent of twinkling stars (Spelbrink et al., 2001 [PubMed 11431692]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ataxin 8; Ataxin8; ATXN 8; C10 orf2; C10orf 2; FLJ21832; mitochondrial twinkle protein; PEO 1; progressive external ophthalmoplegia 1 homolog; Progressive external ophthalmoplegia 1 protein; Progressive external ophthalmoplegia 1 protein homolog; SCA 8; T7 gp4-like protein with intramitochondrial nucleoid localization; T7 helicase-related protein with intramitochondrial nucleoid localization; T7-like mitochondrial DNA helicase; Twinkle mtDNA helicase; Twinkle protein, mitochondrial
Gene Aliases: ATXN8; C10orf2; D19Ertd626e; IOSCA; MTDPS7; PEO; PEO1; PEOA3; PRLTS5; SANDO; SCA8; TWINL; TWNK
UniProt ID: (Human) Q96RR1, (Mouse) Q8CIW5
Entrez Gene ID: (Human) 56652, (Mouse) 226153, (Rat) 309441
Molecular Function:
DNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support