Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: HepG2, DU145, Jurkat, Mouse brain, Rat brain
Immunogen sequence: EDEFSPLKNA DSQNGKDNPT TARHALMSLH HCWVLNAGGH FIDENGSRLP AIPRSATNGK SETITADVNH NLKDANDVPI QCEISPLISY AGEGLESYVA D
Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGX; Antigen X; Protein-pyrophosphorylation enzyme; sperm associated antigen 2; Sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylglucosamine pyrophosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1
Gene Aliases: AA420407; AA437972; AGX; AGX-1; AGX-2; AGX1; AGX2; ESTM38; RGD1561967; SPAG2; UAP1
UniProt ID: (Human) B2R6R8, (Mouse) Q91YN5
Entrez Gene ID: (Human) 6675, (Mouse) 107652, (Rat) 498272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support