Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSSSPLSKKR RVSGPDPKPG SNCSPAQSVL SEVPSVPTNG MAKNGSEADI DEGLYSRQLY VLGHEAMKRL QTSSVLVSGL RGLGVEIAKN IILGGVKAVT LHDQGTAQWA DLSSQFYLRE EDIGKNRAEV SQPRLAELNS YVPVTAYTGP LVEDFLSGFQ VVVLTNTPLE DQLRVGEFCH NRGIKLVVAD TRGLFGQLFC DFGEEMILTD SNGEQPLSAM VSMVTKDNPG VVTCLDEARH GFESGDFVSF SEVQGMVELN GNQPMEIKVL GPYTFSICDT SNFSDYIRGG IVSQVKVPKK ISFKSLVASL AEPDFVVTDF AKFSRPAQLH IGFQALHQFC AQHGRPPRPR NEEDAAELVA LAQAVNARAL PAVQQNNLDE DLIRKLAYVA AGDLAPINAF IGGLAAQEVM KACSGKFMPI MQWLYFDALE CLPEDKEVLT EDKCLQRQNR YDGQVAVFGS DLQEKLGKQK YFLVGAGAIG CELLKNFAMI GLGCGEGGEI IVTDMDTIEK SNLNRQFLFR PWDVTKLKSD TAAAAVRQMN PHIRVTSHQN RVGPDTERIY DDDFFQNLDG VANALDNVDA RMYMDRRCVY YRKPLLESGT LGTKGNVQVV IPFLTESYSS SQDPPEKSIP ICTLKNFPNA IEHTLQWARD EFEGLFKQPA ENVNQYLTDP KFVERTLRLA GTQPLEVLEA VQRSLVLQRP QTWADCVTWA CHHWHTQYSN NIRQLLHNFP PDQLTSSGAP FWSGPKRCPH PLTFDVNNPL HLDYVMAAAN LFAQTYGLTG SQDRAAVATF LQSVQVPEFT PKSGVKIHVS DQELQSANAS VDDSRLEELK ATLPSPDKLP GFKMYPIDFE KDDDSNFHMD FIVAASNLRA ENYDIPSADR HKSKLIAGKI IPAIATTTAA VVGLVCLELY KVVQGHRQLD SYKNGFLNLA LPFFGFSEPL AAPRHQYYNQ EWTLWDRFEV QGLQPNGEEM TLKQFLDYFK TEHKLEITML SQGVSMLYSF FMPAAKLKER LDQPMTEIVS RVSKRKLGRH VRALVLELCC NDESGEDVEV PYVRYTIR
UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1S9 protein; A1S9T and BN75 temperature sensitivity complementing; POC20 centriolar protein homolog; Protein A1S9; testicular secretory protein Li 63; UBA1, ubiquitin-activating enzyme E1 homolog A; Ubiquitin-activating enzyme E1; ubiquitin-like modifier activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1
Gene Aliases: A1S9; A1S9T; A1ST; AMCX1; CFAP124; GXP1; POC20; SMAX2; UBA1; UBA1A; UBE1; UBE1X
UniProt ID: (Human) P22314
Entrez Gene ID: (Human) 7317
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support