Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YDKVELANM NRLFFQPHQA GLSKVQAAEH TLRNINPDVL FEVHNYNITT VENFQHFMDR ISNGGLEEGK PVDLVLSCVD NFEARMTINT ACNELGQTWM ESGVSENAVS GHIQLIIPGE SACFACAPPL VVAANIDEKT LKREGVCAAS LPTTMGVVAG ILVQNVLKFL LNFGTVSFYL GYNAMQDFFP TMSMKPNPQC DDRNCRKQQE EYKKKVAALP KQEVIQEEEE IIHEDNEWGI ELVSEVSEEE LKNFSGPVPD LPEGITVAYT IPKKQEDSVT ELTVEDSGES LEDLMAKMKN M (105-404 aa encoded by BC009737)
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A pseudogene located on chromosome 1 has also been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23251; FLJ23251UBA5; ThiFP1; UBA5, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; Ubiquitin-like modifier-activating enzyme 5; UFM1-activating enzyme
Gene Aliases: 5730525G14Rik; AW240750; THIFP1; UBA5; UBE1DC1
UniProt ID: (Human) Q9GZZ9, (Mouse) Q8VE47
Entrez Gene ID: (Human) 79876, (Mouse) 66663
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support