Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MAESVERLQQ RVQELERELA QERSLQVPRS GDGGGGRVRI EKMSSEVVDS NPYSRLMALK RMGIVSDYEK IRTFAVAIVG VGGVGSVTAE MLTRCGIGKL LLFDYDKVEL ANMNRLFFQP HQAGLSKVQA AEHTLRNINP DVLFEVHNYN ITTVENFQHF MDRISNGGLE EGKPVDLVLS CVDNFEARMT INTACNELGQ TWMESGVSEN AVSGHIQLII PGESACFACA PPLVVAANID EKTLKREGVC AASLPTTMGV VAGILVQNVL KFLLNFGTVS FYLGYNAMQD FFPTMSMKPN PQCDDRNCRK QQEEYKKKVA ALPKQEVIQE EEEIIHEDNE WGIELVSEVS EEELKNFSGP VPDLPEGITV AYTIPKKQED SVTELTVEDS GESLEDLMAK MKNM; Positive Samples: HT-29, A-549, Jurkat, HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus
This gene encodes a member of the E1-like ubiquitin-activating enzyme family. This protein activates ubiquitin-fold modifier 1, a ubiquitin-like post-translational modifier protein, via the formation of a high-energy thioester bond. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A pseudogene located on chromosome 1 has also been identified.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23251; FLJ23251UBA5; ThiFP1; UBA5, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; Ubiquitin-like modifier-activating enzyme 5; UFM1-activating enzyme
Gene Aliases: 5730525G14Rik; AW240750; THIFP1; UBA5; UBE1DC1
UniProt ID: (Human) Q9GZZ9, (Rat) Q5M7A4, (Mouse) Q8VE47
Entrez Gene ID: (Human) 79876, (Rat) 300968, (Mouse) 66663
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support