Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MEGSEPVAAH QGEEASCSSW GTGSTNKNLP IMSTASVEID DALYSRQRYV LGDTAMQKMA KSHVFLSGMG GLGLEIAKNL VLAGIKAVTI HDTEKCQAWD LGTNFFLSED DVVNKRNRAE AVLKHIAELN PYVHVTSSSV PFNETTDLSF LDKYQCVVLT EMKLPLQKKI NDFCRSQCPP IKFISADVHG IWSRLFCDFG DEFEVLDTTG EEPKEIFISN ITQANPGIVT CLENHPHKLE TGQFLTFREI NGMTGLNGSI QQITVISPFS FSIGDTTELE PYLHGGIAVQ VKTPKTVFFE SLERQLKHPK CLIVDFSNPE APLEIHTAML ALDQFQEKYS RKPNVGCQQD SEELLKLATS ISETLEEKVT IEIYGCPNIC LLIHKCSVY
UBA6 (UBE1L2, MOP-4) is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E1-L2; Monocyte protein 4; MOP-4; UBA6, ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme 6; ubiquitin-activating enzyme E1-like 2; Ubiquitin-activating enzyme E1-like protein 2; ubiquitin-like modifier activating enzyme 6; Ubiquitin-like modifier-activating enzyme 6
Gene Aliases: E1-L2; MOP-4; MOP4; UBA6; UBE1L2
UniProt ID: (Human) A0AVT1
Entrez Gene ID: (Human) 55236
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support