Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDALDASKLL DEELYSRQLY VLGSPAMQRI QGARVLVSGL QGLGAEVAKN LVLMGVGSLT LHDPHPTCWS DLAAQFLLSE QDLERSRAEA SQELLAQLNR AVQVVVHTGD ITEDLLLDFQ VVVLTAAKLE EQLKVGTLCH KHGVCFLAAD TRGLVGQLFC DFGEDFTVQD PTEAEPLTAA IQHISQGSPG ILTLRKGANT HYFRDGDLVT FSGIEGMVEL NDCDPRSIHV REDGSLEIGD TTTFSRYLRG GAITEVKRPK TVRHKSLDTA LLQPHVVAQS SQEVHHAHCL HQAFCALHKF QHLHGRPPQP WDPVDAETVV GLARDLEPLK RTEEEPLEEP LDEALVRTVA LSSAGVLSPM VAMLGAVAAQ EVLKAISRKF MPLDQWLYFD ALDCLPEDGE LLPSPEDCAL RGSRYDGQIA VFGAGFQEKL RRQHYLLVGA GAIGCELLKV FALVGLGAGN SGGLTVVDMD HIERSNLSRQ FLFRSQDVGR PKAEVAAAAA RGLNPDLQVI PLTYPLDPTT EHIYGDNFFS RVDGVAAALD SFQARRYVAA RCTHYLKPLL EAGTSGTWGS ATVFMPHVTE AYRAPASAAA SEDAPYPVCT VRYFPSTAEH TLQWARHEFE ELFRLSAETI NHHQQAHTSL ADMDEPQTLT LLKPVLGVLR VRPQNWQDCV AWALGHWKLC FHYGIKQLLR HFPPNKVLED GTPFWSGPKQ CPQPLEFDTN QDTHLLYVLA AANLYAQMHG LPGSQDWTAL RELLKLLPQP DPQQMAPIFA SNLELASASA EFGPEQQKEL NKALEVWSVG PPLKPLMFEK DDDSNFHVDF VVAAASLRCQ NYGIPPVNRA QSKRIVGQII PAIATTTAAV AGLLGLELYK VVSGPRPRSA FRHSYLHLAE NYLIRYMPFA PAIQTFHHLK WTSWDRLKVP AGQPERTLES LLAHLQEQHG LRVRILLHGS ALLYAAGWSP EKQAQHLPLR VTELVQQLTG QAPAPGQRVL VLELSCEGDD EDTAFPPLHY EL
UBE1L (ubiquitin-like modifier-activating enzyme 7) activates ubiquitin by first adenylating with ATP at its C-terminal glycine residue and then linking that residue to the side chain of a cysteine residue in E1. This activation results in an ubiquitin-E1 thioester and free AMP. UBE1L also catalyzes the ISGylation of influenza A virus NS1 protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D8; MGC12713; UBA1, ubiquitin-activating enzyme E1 homolog B; UBA7, ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme 7; Ubiquitin-activating enzyme E1 homolog; ubiquitin-activating enzyme E1-related protein; ubiquitin-activating enzyme-2; ubiquitin-like modifier activating enzyme 7; Ubiquitin-like modifier-activating enzyme 7
Gene Aliases: D8; UBA1B; UBA7; UBE1L; UBE2
UniProt ID: (Human) P41226
Entrez Gene ID: (Human) 7318
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support