Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: MCF7. The target is usually found in the following locations: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Immunogen sequence: PIFSSSEPTS EARIGMGATL DIQRQQRMEL LDRQLMFSQF AQGRRQRQQQ GGMINWNRLF PPLRQRQNVN YQGGRQSEPA APPLEVSEEQ V
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: phosphoglycerate dehydrogenase like 1; Phosphoglycerate dehydrogenase-like protein 1; RP11-178C10.1; UBA domain-containing protein 2; Ubiquitin-associated domain-containing protein 2
Gene Aliases: 1190008A14Rik; AW554959; PHGDHL1; PSEC0110; UBAC2
UniProt ID: (Human) Q8NBM4, (Mouse) Q8R1K1
Entrez Gene ID: (Human) 337867, (Rat) 361094, (Mouse) 68889
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support