Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Immunogen sequence: QQNSTSHPTT TTSWDLKPPT SQSSVLSHLD FKSQPEPSPV LSQLSQRQQH QSQAVTVPPP GLESFPSQAK LRESTPGDSP STVNKLLQLP STTIENISVS VHQPQPKHIK LAKRRIPPAS K
The protein encoded by this gene contains a UBA (ubiquitin associated) domain, which is characteristic of proteins that function in the ubiquitination pathway. This gene may show increased expression in the adrenal gland and lymphatic tissues. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AD-012 protein; RNA polymerase II degradation factor UBAP2; UBAP-2; UBAP2; Ubiquitin-associated protein 2
Gene Aliases: KIAA1491; UBAP-2; UBAP2
UniProt ID: (Human) Q5T6F2
Entrez Gene ID: (Human) 55833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support