Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAQYGHPSPL GMAAREELYS KVTPRRNRQQ RPGTIKHGSA LDVLLSMGFP RARAQKALAS TGGRSVQAAC DWLFSHVGDP FLDDPLPREY VLYLRPTGPL AQKLSDFWQQ SKQICGKNKA HNIFPHITLC QFFMCEDSKV DALGEALQTT VSRWKCKFSA PLPLELYTSS NFIGLFVKED SAEVLKKFAA DFAAEAASKT EVHVEPHKKQ LHVTLAYHFQ ASHLPTLEKL AQNIDVKLGC DWVATIFSRD IRFANHETLQ VIYPYTPQND DELELVPGDF IFMSPMEQTS TSEGWIYGTS LTTGCSGLLP ENYITKADEC STWIFHGSYS ILNTSSSNSL TFGDGVLERR PYEDQGLGET TPLTIICQPM QPLRVNSQPG PQKRCLFVCR HGERMDVVFG KYWLSQCFDA KGRYIRTNLN MPHSLPQRSG GFRDYEKDAP ITVFGCMQAR LVGEALLESN TIIDHVYCSP SLRCVQTAHN ILKGLQQENH LKIRVEPGLF EWTKWVAGST LPAWIPPSEL AAANLSVDTT YRPHIPISKL VVSESYDTYI SRSFQVTKEI ISECKSKGNN ILIVAHASSL EACTCQLQGL SPQNSKDFVQ MVRKIPYLGF CSCEELGETG IWQLTDPPIL PLTHGPTGGF NWRETLLQE
Sts1 encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cbl-interacting protein p70; Cbl-interacting protein Sts-1; MGC15437; nm23-phosphorylated unknown substrate; SH3 domain-containing 70 kDa protein, suppressor of T-cell receptor signaling 1, nm23-phosphorylated unknown substrate; STS-1; Suppressor of T-cell receptor signaling 1; T-cell ubiquitin ligand 2; TULA-2; Tyrosine-protein phosphatase STS1/TULA2; Ubiquitin-associated and SH3 domain-containing protein B
Gene Aliases: KIAA1959; p70; STS-1; STS1; TULA-2; TULA2; UBASH3B
UniProt ID: (Human) Q8TF42
Entrez Gene ID: (Human) 84959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support