Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ACHHWHTQY SNNIRQLLHN FPPDQLTSSG APFWSGPKRC PHPLTFDVNN PLHLDYVMAA ANLFAQTYGL TGSQDRAAVA TFLQSVQVPE FTPKSGVKIH VSDQELQSAN ASVDDSRLEE LKATLPSPDK LPGFKMYPID FEKDDDSNFH MDFIVAASNL RAENYDIPSA DRHKSKLIAG KIIPAIATTT AAVVGLVCLE LYKVVQGHRQ LDSYKNGFLN LALPFFGFSE PLAAPRHQYY NQEWTLWDRF EVQGLQPNGE EMTLKQFLDY FKTEHKLEIT MLSQGVSMLY SFFMPAAKLK ERLDQPMTEI VSRVSKRKLG RHVRALVLEL CCNDESGEDV EVPYVRYTIR (710-1058 aa encoded by BC013041)
UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A1S9 protein; A1S9T and BN75 temperature sensitivity complementing; POC20 centriolar protein homolog; Protein A1S9; testicular secretory protein Li 63; uba1 {ECO:0000312|EMBL:AAH85791.1, ECO:0000312|RGD:1359327}; UBA1, ubiquitin-activating enzyme E1 homolog A; Ubiquitin-activating enzyme E1; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1 {ECO:0000250|UniProtKB:Q02053}; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1 X; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250|UniProtKB:Q02053}
Gene Aliases: A1S9; A1S9T; A1ST; AMCX1; CFAP124; GXP1; POC20; Sbx; SMAX2; UBA1; UBA1A; Ube-1; UBE1; Ube1ax; UBE1X
UniProt ID: (Human) P22314, (Mouse) Q02053, (Rat) Q5U300
Entrez Gene ID: (Human) 7317, (Mouse) 22201, (Rat) 314432
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support