Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MALKRINKEL SDLARDPPAQ CSAGPVGDDM FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TRIYHPNINS NGSICLDILR SQWSPALTIS KVLLSICSLL CDPNPDDPLV PEIARIYKTD RDKYNRISRE WTQKYAM
Ubiquitination is a post-translational modification pathway or is part of the specific protein-degradation pathway by the 26S proteasome. Ubiquitination of a target protein involves multistep enzymatic reaction catalyzed by a cascade of enzymes including Ub-activating enzymes (E1s), Ub-conjugating enzymes (E2s) and Ub ligases (E3s) (PMID: 23542885). UBE2D family, which has 4 members named as UBE2D1/2/3/4, is an E2 ubiquitin-conjugating enzyme family in the ubiquitin-proteasome system. This antibody can recognize all the four members of UBE2D due to the high homology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (E3-independent) E2 ubiquitin-conjugating enzyme D1; (E3-independent) E2 ubiquitin-conjugating enzyme D2; (E3-independent) E2 ubiquitin-conjugating enzyme D3; E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D2; E2 ubiquitin-conjugating enzyme D2B; E2 ubiquitin-conjugating enzyme D3; E2 ubiquitin-conjugating enzyme D4; E2(17)KB 1; E2(17)KB 2; E2(17)KB 2B; E2(17)KB 3; HBUCE1; p53-regulated ubiquitin-conjugating enzyme 1; PAPase; Phosphoarginine phosphatase; PRO2116; SFT; Stimulator of Fe transport; ubc2e ubiquitin conjugating enzyme; UBC4/5 homolog; UbcH5; UBE2D1/UBE2D3/UBE2D3/UBE2D4; Ubiquitin carrier protein D1; Ubiquitin carrier protein D2; Ubiquitin carrier protein D2B; Ubiquitin carrier protein D3; Ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2D 1; ubiquitin conjugating enzyme E2D 2; ubiquitin conjugating enzyme E2D 3; ubiquitin conjugating enzyme E2D 4 (putative); Ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2 D2B; Ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2 D4; Ubiquitin-conjugating enzyme E2(17)KB 1; Ubiquitin-conjugating enzyme E2(17)KB 2; Ubiquitin-conjugating enzyme E2(17)KB 2B; Ubiquitin-conjugating enzyme E2(17)KB 3; Ubiquitin-conjugating enzyme E2-17 kDa 1; Ubiquitin-conjugating enzyme E2-17 kDa 2; Ubiquitin-conjugating enzyme E2-17 kDa 2B; Ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme HBUCE1; Ubiquitin-protein ligase D1; Ubiquitin-protein ligase D2; Ubiquitin-protein ligase D2B; Ubiquitin-protein ligase D3; Ubiquitin-protein ligase D4
Gene Aliases: 1100001F19Rik; 9430029A22Rik; AA414951; E2(17)KB1; E2(17)KB2; E2(17)KB3; E217kB; HBUCE1; PUBC1; RGD69425; SFT; Ubc2e; UBC4; UBC4/5; UBC5A; UBC5B; UBC5C; UBCH4; UBCH5; UBCH5A; UBCH5B; UBCH5C; UBCH5D; UBE2D1; UBE2D2; Ube2d2b; UBE2D3; UBE2D4
UniProt ID: (Human) P51668, (Human) P62837, (Human) P61077, (Human) Q9Y2X8, (Mouse) P61080, (Rat) D3ZDK2, (Mouse) P61079, (Rat) P61078, (Rat) P70711
Entrez Gene ID: (Human) 7321, (Human) 7322, (Human) 7323, (Human) 51619, (Mouse) 216080, (Rat) 361831, (Rat) 641452, (Mouse) 66105, (Rat) 81920, (Rat) 79435
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support