Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MIKLFSLKQQ KKEEESAGGT KGSSKKASAA QLRIQKDINE LNLPKTCDIS FSDPDDLLNF KLVICPDEGF YKSGKFVFSF KVGQGYPHDP PKVKCETMVY HPNIDLEGNV CLNILREDWK PVLTINSIIY GLQYLFLEPN PEDPLNKEAA EVLQNNRRLF EQNVQRSMRG GYIGSTYFER CLK
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEDD8 carrier protein; NEDD8 protein ligase; NEDD8-conjugating enzyme Ubc12; ubiquitin carrier protein M; ubiquitin conjugating enzyme E2M; Ubiquitin-conjugating enzyme E2 M; ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast); ubiquitin-protein ligase M; yeast UBC12 homolog
Gene Aliases: hUbc12; UBC-RS2; UBC12; UBE2M
UniProt ID: (Human) P61081
Entrez Gene ID: (Human) 9040
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support