Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EPPPGMFVV PDTVDMTKIH ALITGPFDTP YEGGFFLFVF RCPPDYPIHP PRVKLMTTGN NTVRFNPNFY RNGKVCLSIL GTWTGPAWSP AQSISSVLIS IQSLMTENPY HNEPGFEQER HPGDSKNYNE CIRHETIRVA VCDMMEGKCP CPEPLRGVME KSFLEYYDFY EVACKDRLHL QGQTMQDPFG EKRGHFDYQS LLMRLGLIRQ KVLERLHNEN AEMDSDSSSS GTETDLHGSL RV (6-246 aa encoded by BC015169)
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 ubiquitin-conjugating enzyme Z; Uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2Z; Ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); Ubiquitin-protein ligase Z; Use1
Gene Aliases: AW049199; C030047H17Rik; D11Moh35; HOYS7; RGD1308347; UBE2Z; USE1
UniProt ID: (Human) Q9H832, (Rat) Q3B7D1, (Mouse) Q3UE37
Entrez Gene ID: (Human) 65264, (Rat) 303478, (Mouse) 268470
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support