Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FKLLAEKVEE IVAKNARAEI DYSDAPDEFR DPLMDTLMTD PVRLPSGTIM DRSIILRHLL NSPTDPFNRQ TLTESMLEPV PELKEQIQAW MREKQNSDH
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homologous to yeast UFD2; Homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; RING-type E3 ubiquitin transferase E4 B; Ubiquitin conjugation factor E4 B; Ubiquitin fusion degradation protein 2; ubiquitin-fusion degradation protein 2; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); UFD2A-III/UBE4B-III splice isoform
Gene Aliases: E4; HDNB1; KIAA0684; UBE4B; UBOX3; UFD2; UFD2A
UniProt ID: (Human) O95155
Entrez Gene ID: (Human) 10277
Molecular Function: ubiquitin-protein ligase protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support