Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
PA1-4378 detects the TERE1 protein in human samples.
PA1-4378 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~37 kDa protein representing TERE1 in urothelial carcinoma.
The PA1- 4378 immunogen is a synthetic peptide corresponding to residues P(32) E Q D R L P Q R S W R Q K(45) of human TERE1. There is 85% sequence identity to mouse and rat.
A novel gene designated TERE1, has a significant effect on the growth regulation in bladder cancer. The TERE1 gene maps to a chromosome locus that has been identified by loss of heterozygosity studies as a site of a putative tumor suppressor gene or genes for multiple tumor types including prostate carcinoma. Data suggest that TERE1 may be significant in prostate cancer growth regulation and the down regulation or absence of TERE1 may be an important component of the phenotype of advanced disease. Data also suggest a potential role for this gene product in the progression of bladder cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP4-796F18.1; transitional epithelia response protein; Transitional epithelial response protein 1; UbiA prenyltransferase domain-containing protein 1
Gene Aliases: SCCD; TERE1; UBIAD1
UniProt ID: (Human) Q9Y5Z9
Entrez Gene ID: (Human) 29914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support