Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • UBOX5 Antibodies

            Proteintech

            UBOX5 Monoclonal Antibody (2A7A4)

            View all (21) UBOX5 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite UBOX5 Monoclonal Antibody (2A7A4)

            • Antibody Testing Data (2)
            Application
            UBOX5 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            UBOX5 Antibody (68422-1-IG) in WB

            Various lysates were subjected to SDS PAGE followed by western blot with 68422-1-Ig (UBOX5 antibody) at dilution of 1:5,000 incubated at room temperature for 1.5 hours. The membrane was stripped and reblotted with HRP-conjugated GAPDH Monoclonal antibody (HRP-60004) as loading control.
            View Product

            UBOX5 Antibody in Western Blot (WB)
            UBOX5 Antibody in Western Blot (WB)

            Product Details

            68422-1-IG

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:2,000-1:10,000
            -

            ELISA (ELISA)

            Assay-dependent
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG1

            Class

            Monoclonal

            Type

            Antibody

            Clone

            2A7A4

            Immunogen

            UBOX5 fusion protein Ag6573
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice

            Product Specific Information

            Immunogen sequence: QPAKTCSQEV IDSILLVTSE NLPQDVALQA PALPMESDCD PGDQPESQQA PSSLQKLAEI IQDVPEEFLD PITLEIMPCP MLLPSGKVID QSTLEKCNRS EATWGRVPSD PFTGVAFTPH SQPLPHPSLK ARIDHFLLQH SIPGCHLLGR AQTALAVIPS SIVLPSQKRK IEQAEHVPDS NFGVNASCFS ATSPLVLPTT SEHTAKKMKA TNEPSLTHMD CSTGPLSHEQ KLSQSLEIAL ASTLGSMPSF TARLTRGQLQ HLGTRGSNTS WRPGTGSEQP GSILGPECAS CKRVFSPYFK KEPVYQLPCG HLLCRPCLGE KQRSLPMTCT ACQRPVASQD VLRVHF

            Target Information

            UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: hUIP5; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; U-box domain-containing protein 5; UbcM4-interacting protein 5; ubiquitin conjugating enzyme 7 interacting protein 5; Ubiquitin-conjugating enzyme 7-interacting protein 5

            View more View less

            Gene Aliases: hUIP5; KIAA0860; RNF37; UBCE7IP5; UBOX5; UIP5

            View more View less

            UniProt ID: (Human) O94941

            View more View less

            Entrez Gene ID: (Human) 22888

            View more View less

            Function(s)

            protein binding zinc ion binding ligase activity ubiquitin protein ligase binding ubiquitin-ubiquitin ligase activity

            Process(es)

            protein polyubiquitination
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0