Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSRVHQPTG MNQAPVELAH FVGSDAPKVH TQNLEVSHPE CKAQMLENPS IQRLLSNMEF MWQFISEHLD TQQLMQQNPE VSRLLLDNSE ILLQTLELAR NLAMIQEIMQ IQQPSQNLEY PLNPQPYLGL ETMPGGNNAL GQNYVDINDQ MLNSMQDPFG GNPFTALLAG QVLEQVQSSP PPPPPSQEQQ DQLTQHPATR VIYNSSGGFS SNTSANDTLN KVNHTSKANT AMISTKGQSH ICATRQPAGI PALPSIELTQ QLQEEYKDAT VSLSSSRQTL KGDLQLSDEQ SSSQITGGMM QLLMNNPYLA AQIMLFTSMP QLSEEWRQQL PTFLQQTQIS DLLSA (132-475 aa encoded by BC012183)
UBQLNL is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ubiquilin-like protein
Gene Aliases: 4922504M18Rik; Gm490; UBQLNL
UniProt ID: (Human) Q8IYU4, (Mouse) Q14DL0
Entrez Gene ID: (Human) 143630, (Mouse) 244179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support