Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The mammalian NUMB homolog plays a role in the determination of cell fate during development and binds with a variety of proteins, including Eps15, LNX1 and Notch 1. NumbL (NUMB-like protein), also known as Numb-R, NBL, CAG3A, CTG3a, NUMBLIKE or TNRC23, is a 609 amino acid cytoplasmic protein that, like NUMB, is thought to play a role in cell fate. Expressed at high levels in developing brain tissue, NumbL contains one PID (phosphotyrosine interaction domain) and plays an important role in neuronal differentiation, possibly associating with Eps15 and Notch 1. In mice, deletion of the NumbL gene is associated with early embryonic death, suggesting an essential role for NumbL in early development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase UBR2; N-recognin-2; RING-type E3 ubiquitin transferase UBR2; ubiquitin ligase E3 alpha-II; Ubiquitin-protein ligase E3-alpha-2; Ubiquitin-protein ligase E3-alpha-II
Gene Aliases: 9930021A08Rik; AI462103; AW540746; bA49A4.1; C6orf133; dJ242G1.1; dJ392M17.3; E130209G04Rik; KIAA0349; mKIAA0349; UBR2
UniProt ID: (Human) Q8IWV8, (Mouse) Q6WKZ8
Entrez Gene ID: (Human) 23304, (Mouse) 224826, (Rat) 363188
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support