Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTGNAGEWC LMESDPGVFT ELIKGFGCRG AQVEEIWSLE PENFEKLKPV HGLIFLFKWQ PGEEPAGSVV QDSRLDTIFF AKQVINNACA TQAIVSVLLN CTHQDVHLGE TLSEFKEFSQ SFDAAMKGLA LSNSDVIRQV HNSFARQQMF EFDTKTSAKE EDAFHFVSYV PVNGRLYELD GLREGPIDLG ACNQDDWISA VRPVIEKRIQ KYSEGEIRFN LMAIVSDRKM IYEQKIAELQ RQLAEEPMDT DQGNSMLSAI QSEVAKNQML IEEEVQKLKR YKIENIRRKH NYLPFIMELL KTLAEHQQLI PLVEKFEKHF EKTLLGK (1-326 aa encoded by BC025369)
UCH-L5 (ubiquitin carboxyl-terminal hydrolase isozyme L5), also known as UCH37, is a 329 amino acid protein that functions to edit polyubiquinated protein substrates. Since UCH-L5 has the potential to rescue ubiquinated proteins, including oncogenic proteins, from proteasomal degradation, it is likely that deregulation of UCH-L5 may affect tumor growth. Through associations with Smad7, UHC-L5 can dramatically upregulate TGFβ-dependent gene expression by de-ubiquinating and stabilizing TGFβ RI. Also, since overexpression of UCH-L5 and other deubiquitinating enzymes has been observed in many cancer cell lines, inhibition of these proteins may be of some interest in designing therapies for cancer treatment. There are four isoforms of UCH-L5 that exist as a result of alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: INO80 complex subunit R; ubiquitin C-terminal hydrolase 37; Ubiquitin C-terminal hydrolase UCH37; ubiquitin carboxyl-terminal esterase L5; Ubiquitin carboxyl-terminal hydrolase isozyme L5; ubiquitin carboxyl-terminal hydrolase L5; Ubiquitin thioesterase L5; UCH-L5
Gene Aliases: 5830413B11Rik; AD-019; CGI-70; INO80R; UCH-L5; UCH37; UCHL5
UniProt ID: (Human) Q9Y5K5, (Mouse) Q9WUP7
Entrez Gene ID: (Human) 51377, (Rat) 360853, (Mouse) 56207
Molecular Function: cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support