Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytidine monophosphokinase 2; Testis-specific protein TSA903; UCK 2; uridine kinase; uridine monophosphate kinase; Uridine monophosphokinase 2; Uridine-cytidine kinase 2
Gene Aliases: TSA903; UCK2; UK; UMPK
UniProt ID: (Human) Q9BZX2
Entrez Gene ID: (Human) 7371
Molecular Function: nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support