Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVGFKATDVP PTATVKFLGA GTAACIADLI TFPLGTAKVR LQIQGESQGP VRATASAQYR GVMGTILTMV RTEGPRSLYN GLVAGLQRQM SFASVRIGLY DSVKQFYTKG SEHASIGSRL LAGSTTGALA VAVAQPTDVV KVRFQAQARA GGGRRYQSTV NAYKTIAREE GFRGLWKGTS PNVARNAIVN CAELVTYDLI KDALLKANLM TDDLPCHFTS AFGAGFCTTV IASPVDVVKT RYMNSALGQY SSAGHCALTM LQKEGPRAFY KGFMPSFLRL GSWNVVMFVT YEQLKRALMA ACTSREAPF
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dicarboxylate carrier SLC25A8; Mitochondrial uncoupling protein 2; Solute carrier family 25 member 8; UCP 2; UCPH; uncoupling protein; uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases: BMIQ4; SLC25A8; UCP2; UCPH
UniProt ID: (Human) P55851
Entrez Gene ID: (Human) 7351
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support