FIGURE: 1 / 1
The protein encoded by this gene converts UDP-glucose to UDP-glucuronate and thereby participates in the biosynthesis of glycosaminoglycans such as hyaluronan, chondroitin sulfate, and heparan sulfate. These glycosylated compounds are common components of the extracellular matrix and likely play roles in signal transduction, cell migration, and cancer growth and metastasis. The expression of this gene is up-regulated by transforming growth factor beta and down-regulated by hypoxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP UGD; UDP-Glc dehydrogenase; UDP-glucose 6-dehydrogenase; UDP-glucose dehydrogenase; uridine diphospho-glucose dehydrogenase
Gene Aliases: GDH; UDP-GlcDH; UDPGDH; UGD; UGDH
UniProt ID: (Human) O60701
Entrez Gene ID: (Human) 7358
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support