Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: EENFFTLTTY AISWTQDEFD RLLLGHTQSF FETEHLLMKF SRRMAIMNNM
Sequence homology: Human: 100%; Rat: 83%
This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP glucuronosyltransferase 1 family, polypeptide A5; UDP glycosyltransferase 1 family, polypeptide A5; UDP-glucuronosyltransferase 1 family polypeptide A5s; UDP-glucuronosyltransferase 1-5; UDP-glucuronosyltransferase 1-E; UDP-glucuronosyltransferase 1A5; UDPGT 1-5; UGT-1E; UGT1*5; UGT1A5
Gene Aliases: GNT1; UDPGT; UDPGT 1-5; UGT1; UGT1A5; UGT1E
UniProt ID: (Human) P35504
Entrez Gene ID: (Human) 54579
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support