Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QVISWLAPE DHQREFKKSF DFFLEETLGG RGKFENLLNV LEYLALQCSH FLNRKD (75-129 aa encoded by BC103925)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP glucuronosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDPGT 3A2
Gene Aliases: AI313915; PSEC0073; UGT3A2; UNQ842/PRO1780
UniProt ID: (Human) Q3SY77, (Mouse) Q8JZZ0
Entrez Gene ID: (Human) 167127, (Mouse) 223337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support