Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DQSSQPVFE NVNVKSFDRC TGHSAEHTQC GKPQSSQGIV EETSEEGNSV PASQSVAALT SKRSLVLMPE SSAEEITVCP ETQLSSSETF DLEREVSPGS RDILDGVRII MADKEVGNKE DAEKEVAIST FSSSNQVSCP LCDQCFPPTK IERHAMYCNG LMEEDTVLTR RQKEAKTKSD SGTAAQTSLD IDKNEKCYLC KSLVPFREYQ CHVDSCLQLA KADQGDGPEG SGRACSTVEG KWQQRLKNPK EKGHSEGRLL SFLEQSEHKT SDADIKSSET GAFRVPSPGM EEAGCSREMQ SSFTRRDLNE SPVKSFVSIS EATDCLVDFK KQVTVQPGSR TRTKAGRGRR RKF (368-719 aa encoded by BC032561)
RAP80 was initially identified as zinc-finger containing nuclear protein that is highly expressed in testis and interacts with the retinoid-related testis-associated receptor (RTR). Later experiments revealed that RAP80 is recruited by the Coiled-coil domain 98 (CCDC98) protein to the breast cancer-1 protein BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both RAP80 and CCDC98 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either RAP80 or CCDC98 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that RAP80 and CCDC98 control only part of the DNA damage response role of BRCA1. At least four isoforms of RAP80 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRCA1-A complex subunit RAP80; Receptor-associated protein 80; retinoid X receptor interacting protein 110; Retinoid X receptor-interacting protein 110; Ubiquitin interaction motif-containing protein 1
Gene Aliases: 9430016E08Rik; D330018D10Rik; D630032M02Rik; RAP80; RIP110; RXRIP110; UIMC1; X2HRIP110
UniProt ID: (Human) Q96RL1, (Mouse) Q5U5Q9
Entrez Gene ID: (Human) 51720, (Mouse) 20184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support