Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
MA5-31948 has been tested in indirect ELISA.
ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC:2.7.11.1; Serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; Unc-51 like kinase 2; Unc-51-like kinase 2
Gene Aliases: A830085I22Rik; ATG1B; AU015340; KIAA0623; mKIAA0623; ULK2; Unc51.2
UniProt ID: (Human) Q8IYT8, (Mouse) Q9QY01
Entrez Gene ID: (Human) 9706, (Mouse) 29869, (Rat) 303206
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support