Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
UMODL1 gene ontology annotations related to this gene include adipose tissue development; cellular response to gonadotropin-releasing hormone; multicellular organismal reproductive process; regulation of apoptotic process; regulation of gene expression; regulation of ovarian follicle development; single fertilization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Olfactorin; Uromodulin-like 1
Gene Aliases: D17Ertd488e; UMODL1
UniProt ID: (Human) Q5DID0, (Mouse) Q5DID3
Entrez Gene ID: (Human) 89766, (Mouse) 52020
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support