Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MKVKKGGGGA GTATESAPGP SGQSVAPIPQ PPAESESGSE SEPDAGPGPR PGPLQRKQPI GPEDVLGLQR ITGDYLCSPE ENIYKIDFVR FKIRDMDSGT VLFEIKKPPV SERLPINRRD LDPNAGRFVR YQFTPAFLRL RQVGATVEFT VGDKPVNNFR MIERHYFRNQ LLKSFDFHFG FCIPSSKNTC EHIYDFPPLS EELISEMIRH PYETQSDSFY FVDDRLVMHN KADYSYSGTP
This gene is specifically expressed in the photoreceptors in the retina. The encoded product shares strong homology with the C. elegans unc119 protein and it can functionally complement the C. elegans unc119 mutation. It has been localized to the photoreceptor synapses in the outer plexiform layer of the retina, and suggested to play a role in the mechanism of photoreceptor neurotransmitter release through the synaptic vesicle cycle. Two transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hRG4; POC7 centriolar protein homolog A; Protein unc-119 homolog A; Retinal protein 4; unc-119 homolog
Gene Aliases: HRG4; IMD13; POC7; POC7A; RG4; UNC119
UniProt ID: (Human) Q13432
Entrez Gene ID: (Human) 9094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support