Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ATLLSHPQQR PPFLRQAIKI RRRRVRDLQD PPPQMAPEIQ PPSHHFSPEQ RALLYEDALY TVLHRLGHPE PNHVTEASEL LRYLQEAFHV EPEEHQQTL
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: likely ortholog of H. sapiens unc-13 homolog D (UNC13D); munc 13-4; Munc13-4; Protein unc-13 homolog D
Gene Aliases: 2610108D09Rik; FHL3; HLH3; HPLH3; Jinx; mFLJ00067; Munc13-4; UNC13D
UniProt ID: (Human) Q70J99, (Mouse) B2RUP2
Entrez Gene ID: (Human) 201294, (Mouse) 70450
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support