Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: GTDSGSEVLP DSFPSAPAEP LPYFLQEPQD AYIVKNKPVE LRCRAFPATQ IYFKCNGEWV SQNDHVTQEG LDEATGLRVR EVQIEVSRQQ VEELFGLEDY
Receptor for netrin required for axon guidance. Mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Mediates apoptosis by activating DAPK1. In the absence of NTN1, activates DAPK1 by reducing its autoinhibitory phosphorylation at Ser-308 thereby increasing its catalytic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C. elegans; Netrin receptor UNC5B; p53-regulated receptor for death and life protein 1; p53RDL1; Protein unc-5 homolog 2; Protein unc-5 homolog B; transmembrane receptor Unc5H2; unc-5 homolog 2; unc-5 homolog B
Gene Aliases: P53RDL1; UNC5B; UNC5H2; UNQ1883/PRO4326
UniProt ID: (Human) Q8IZJ1
Entrez Gene ID: (Human) 219699
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support