Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: TDNGEALPES IPSAPGTLPH FIEEPDDAYI IKSNPIALRC KARPAMQIFF KCNGEWVHQN EHVSEETLDE SSGLKVREVF INVTRQQVED FHGPEDYWCQ
UNC5D, also named as KIAA1777 and UNC5H4, belongs to the unc-5 family. It is a receptor for netrin. It may be involved in axon guidance by mediating axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16019; KIAA1777Unc5h4; Netrin receptor UNC5D; netrin receptor Unc5h4; Protein unc-5 homolog 4; Protein unc-5 homolog D; unc-5 homolog 4; unc-5 homolog D
Gene Aliases: KIAA1777; PRO34692; UNC5D; UNC5H4; UNQ6012/PRO34692
UniProt ID: (Human) Q6UXZ4
Entrez Gene ID: (Human) 137970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support