Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDFSRLHMYS PPQCVPENTG YTYALSSSYS SDALDFETEH KLDPVFDSPR MSRRSLRLAT TACTLGDGEA VGADSGTSSA VSLKNRAART TKQRRSTNKS AFSINHVSRQ VTSSGVSHGG TVSLQDAVTR RPPVLDESWI REQTTVDHFW GLDDDGDLKG GNKAAIQGNG DVGAAAATAH NGFSCSNCSM LSERKDVLTA HPAPPGPVSR VYSRDRNQKC KSQSFKTQKK VCFPNLIFPF CKSQCLHYLS WRLKIIP
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein unc-84 homolog A; Sad1 unc-84 domain protein 1; Sad1/unc-84 protein-like 1; SUN domain-containing protein 1; unc-84 homolog A
Gene Aliases: KIAA0810; SUN1; UNC84A
UniProt ID: (Human) O94901
Entrez Gene ID: (Human) 23353
Molecular Function:
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support