Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSVSKAAAA ALSGSPPQTE KPTHYRYLKE FRTEQCPLFS QHKCAQHRPF TCFHWHFLNQ RRRRPLRRRD GTFNYSPDVY CSKYNEATGV CPDGDECPYL HRTTGDTERK YHLRYYKTGT CIHETDARGH CVKNGLHCAF AHGPLDLRPP VCDVRELQAQ EALQNGQLGG GEGVPDLQPG VLASQAMIEK ILSEDPRWQD ANFVLGSYKT EQCPKPPRLC RQGYACPHYH NSRDRRRNPR RFQYSWQLGR RVLRLSPRAN NPRVALPRVH TGPSSTA
May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Putative E3 ubiquitin-protein ligase UNKL; RING finger protein unkempt-like; unkempt family zinc finger-like; unkempt homolog-like; Zinc finger CCCH domain-containing protein 5-like
Gene Aliases: C16orf28; UNKL; ZC3H5L; ZC3HDC5L
UniProt ID: (Human) Q9H9P5
Entrez Gene ID: (Human) 64718
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support