Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTCCSQVPPR RRPSLALSPR LDCNGLNGVP GSIWDFVSGS FSPSPSPILS AGPPSSSSAS PNGAELARVR RQLDEAKRKI RQWEESWQQV KQVCDAWQRE AQEAKERARV ADSDRQLALQ KKEEVEAQVK QLQEELEGLG VASTLPGLRG CGDIGTIPLP KLHSLQSQLR LDLEAVDGVI FQLRAKQCVA CRERAHGAVL RPCQHHILCE PCAATAPECP YCKGQPLQW
May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Putative E3 ubiquitin-protein ligase UNKL; RING finger protein unkempt-like; unkempt family zinc finger-like; unkempt homolog-like; Zinc finger CCCH domain-containing protein 5-like
Gene Aliases: C16orf28; UNKL; ZC3H5L; ZC3HDC5L
UniProt ID: (Human) Q9H9P5
Entrez Gene ID: (Human) 64718
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support