Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
UPP1 catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:7488099). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPase 1; urdPase 1; Uridine phosphorylase 1
Gene Aliases: AI325217; UDRPASE; UP; UPASE; UPP; UPP1
UniProt ID: (Human) Q16831, (Mouse) P52624
Entrez Gene ID: (Human) 7378, (Mouse) 22271, (Rat) 289801
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support