Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
FIGURE: 1 / 1
Catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:12849978, PubMed:21855639). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis (Probable). Shows broad substrate specificity and accepts uridine, deoxyuridine, and thymidine as well as the two pyrimidine nucleoside analogs 5-fluorouridine and 5-fluoro-2(')-deoxyuridine as substrates (PubMed:12849978). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver-specific uridine phosphorylase; UPase 2; urdPase 2; Uridine phosphorylase 2; uridine phosphorylase-2
Gene Aliases: UDRPASE2; UP2; UPASE2; UPP2
UniProt ID: (Human) O95045
Entrez Gene ID: (Human) 151531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support