Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain
Predicted Reactivity: Rhesus Monkey (97%), Chimpanzee (100%), Bovine (88%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 1; Core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; ubiquinol-cytochrome c reductase core protein 1; Ubiquinol-cytochrome-c reductase complex core protein 1
Gene Aliases: 1110032G10Rik; D3S3191; QCR1; UQCR1; UQCRC1
UniProt ID: (Human) P31930, (Rat) Q68FY0, (Mouse) Q9CZ13
Entrez Gene ID: (Human) 7384, (Rat) 301011, (Mouse) 22273
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support