Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IEAVGGKLSV TATRENMAYT VECLRGDVDI LMEFLLNVTT APEFRRWEVA DLQPQLKIDK AVAFQNPQTH VIENLHAAAY RNALANPLYC PDYRIGKVTS EELHYFVQNH FTSARMALIG LGVSHPVLKQ VAEQFLNMRG GLGLSGAKAN YRGGEIREQN GDSLVHAAFV AESAVAGSAE ANAFSVLQHV LGAGPHVKRG SNTTSHLHQA VAKATQQPFD VSAFNASYSD SGLFGIYTIS QATAAGDVIK AAYNQVKTIA QGNLSNTDVQ AAKNKLKAGY LMSVESSECF LEEVGSQALV AGSYMPPSTV LQQIDSVANA DIINAAKKFV SGQKSMAASG NLGHTPFVDE L
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 2; Core protein II; Cytochrome b-c1 complex subunit 2, mitochondrial; cytochrome bc-1 complex core protein II; U MC3DN5; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; Ubiquinol-cytochrome-c reductase complex core protein 2
Gene Aliases: 1500004O06Rik; MC3DN5; QCR2; UQCR2; UQCRC2
UniProt ID: (Human) P22695, (Mouse) Q9DB77, (Rat) P32551
Entrez Gene ID: (Human) 7385, (Mouse) 67003, (Rat) 293448
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support